Learn More
Aviva Systems Biology Corp GORASP1 Antibody - N-terminal region (ARP57680_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57680_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GORASP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GORASP1. Alternate names: FLJ23443, GOLPH5, GRASP65, MGC118894, MGC118897, P65ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-63028