Learn More
Aviva Systems Biology Corp GORASP2 Antibody - C-terminal region (ARP62077_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62077_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GORASP2. It was validated on Western Blot. . Alternate names: DKFZp434D156, FLJ13139, GOLPH6, GRASP55, GRS2, p59ApplicationWB. Species reactivity: Human, Rabbit, Rat
Catalog No. 50-287-559