Learn More
Medchemexpress LLC Goserelin (acetate) | 145781-92-6 | 99.9% | 1329.46 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY13673A100MG
Encompass_Preferred
Goserelin acetate (ICI-118630 acetate) is a decapeptide analogue of gonadotropin-releasing hormone (GnRH/LHRH) that acts as a GnRH agonist. It is used for the research of breast cancer, epithelial ovarian cancer, and prostate cancer.
  • Functions as a GnRH agonist.
  • Promotes EOC cell apoptosis.
  • Regulates expression of human apoptosis-related genes.
  • Increases apoptotic cells in subcutaneous xenograft tumors.
Catalog No. 50-003-50942