Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Goserelin (acetate) | 145781-92-6 | MFCD01747329 | 99.9% | C61H88N18O16 | 10MG
SDP

Catalog No. 5000259745
Encompass_Preferred

Goserelin (acetate) is a synthetic decapeptide analog of gonadotropin-releasing hormone (GnRH) used as a research-grade GnRH agonist for in vitro and in vivo studies of hormone-dependent cancers, reproductive biology, and endocrine signaling. Provided as the acetate salt with high analytical purity, it is supplied as a white to off-white solid and intended for research use only.

  • Purity 99.9% (HPLC).
  • Molecular formula C61H88N18O16 and molecular weight 1329.46.
  • White to off-white solid suitable for analytical and biological assays.
  • Supplied in small vial sizes suitable for dosing and assay development.
  • Stable when stored sealed, away from moisture; powder: -80°C (up to 2 years), -20°C (up to 1 year).
  • Acetate salt form for standard biochemical handling.
  • For research use only; not for human or veterinary use.

Catalog No. 50-002-59745 Supplier Medchemexpress LLC Supplier No. HY13673A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goserelin (acetate) is a synthetic decapeptide analog of gonadotropin-releasing hormone (GnRH) used as a research-grade GnRH agonist for in vitro and in vivo studies of hormone-dependent cancers, reproductive biology, and endocrine signaling. Provided as the acetate salt with high analytical purity, it is supplied as a white to off-white solid and intended for research use only.

  • Purity 99.9% (HPLC).
  • Molecular formula C61H88N18O16 and molecular weight 1329.46.
  • White to off-white solid suitable for analytical and biological assays.
  • Supplied in small vial sizes suitable for dosing and assay development.
  • Stable when stored sealed, away from moisture; powder: -80°C (up to 2 years), -20°C (up to 1 year).
  • Acetate salt form for standard biochemical handling.
  • For research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.