Learn More
Medchemexpress LLC Goserelin acetate (ICI-118630 acetate) | 145781-92-6 | MFCD00867894 | 99.9% | 1329.46 g/mol | C61H88N18O16 | 5 MG
SDP
Catalog No. 5000222611
Encompass_Preferred
Goserelin acetate is a synthetic GnRH (gonadotropin-releasing hormone) agonist peptide supplied for research use in studies of hormone-dependent cancers and reproductive biology. The acetate salt is formulated for laboratory assays and preclinical work where a defined GnRH receptor agonist is required.
  • High reported purity (99.87%) suitable for research applications.
  • Well-characterized molecular formula and weight for identity confirmation.
  • Supplied in small milligram quantities for laboratory experiments.
  • Appropriate for in vitro and preclinical cancer biology studies.
  • Datasheet and handling instructions available for proper storage and use.
Catalog No. 50-002-22611 Supplier Medchemexpress LLC Supplier No. HY13673A5MG
Goserelin acetate is a synthetic GnRH (gonadotropin-releasing hormone) agonist peptide supplied for research use in studies of hormone-dependent cancers and reproductive biology. The acetate salt is formulated for laboratory assays and preclinical work where a defined GnRH receptor agonist is required.
  • High reported purity (99.87%) suitable for research applications.
  • Well-characterized molecular formula and weight for identity confirmation.
  • Supplied in small milligram quantities for laboratory experiments.
  • Appropriate for in vitro and preclinical cancer biology studies.
  • Datasheet and handling instructions available for proper storage and use.