Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Assembled HDPE Round Bottles, Red Cap with Wad
GREENER_CHOICE

Catalog No. 07202544 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
150 mL
250 mL
500 mL
1 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
07-202-544 50 mL
07-202-547 100 mL
07-202-549 150 mL
07-202-545 250 mL
07-202-546 500 mL
07-202-548 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 07-202-544 Supplier Corning™ Supplier No. LR50A18
Add to Cart
Add to Cart
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'

Easier to handle, require less space (13-20%) on the shelf and are ideal for sampling and storage. Corning™ HDPE Round Bottles are lightweight and break resistant

  • Break-resistant and translucent high-density polyethylene (HDPE)
  • HDPE cap

Compliance

Food contact compliant

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 50 mL
Color Red
Description Sampling and Storage Bottle
For Use With (Application) Clinical, Industry, Microbiology
Material HDPE
Length (Metric) 77 mm
No. per Case 600/Cs.
Sterility Sterile
Closure Color Red
Certifications/Compliance Raw material is compliant with regulation EC N°1935/2004 Sterility Assurance Level based on ISO 11137 : STERILE SAL 10-3
Product Type Storage Bottle
Closure Liner Wad
Closure Type Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.