Learn More
Medchemexpress LLC HY-13407 100mg Medchemexpress, Gossypol CAS:303-45-7 Purity:>98%
SDP
Catalog No. 502081355
Encompass_Preferred
Medchemexpress, HY-13407 100mg Gossypol CAS:303-45-7 Gossypol, a natural product isolated from cottonseeds and roots, binds to Bcl-xL protein and Bcl-2 protein with Kis of 0.5-0.6 μM and 0.2-0.3 mM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1355 Supplier Medchemexpress LLC Supplier No. HY13407100MG
Medchemexpress, HY-13407 100mg Gossypol CAS:303-45-7 Gossypol, a natural product isolated from cottonseeds and roots, binds to Bcl-xL protein and Bcl-2 protein with Kis of 0.5-0.6 μM and 0.2-0.3 mM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.