Learn More
Medchemexpress LLC Gossypol (acetic acid) | 12542-36-8 | 578.61 | 100 MG
SDP
Catalog No. 5000386386
Encompass_Preferred
Gossypol acetic acid ((±)-Gossypol-acetic acid) binds to Bcl-xL protein and Bcl-2 protein with Kis of 0.5-0.6 μM and 0.2-0.3 mM, respectively. It is harmful if swallowed and suspected of causing cancer. Proper handling and storage are crucial, including avoiding inhalation and contact with eyes and skin, using appropriate ventilation, and storing in a cool, well-ventilated area away from direct sunlight and ignition sources.
  • Binds to Bcl-xL protein and Bcl-2 protein
  • Harmful if swallowed
  • Suspected of causing cancer
  • Avoid inhalation and contact with eyes and skin
  • Use in areas with appropriate exhaust ventilation
  • Keep container tightly sealed in a cool, well-ventilated area
  • Keep away from direct sunlight and sources of ignition
Catalog No. 50-003-86386 Supplier Medchemexpress LLC Supplier No. HY17510100MG
Gossypol acetic acid ((±)-Gossypol-acetic acid) binds to Bcl-xL protein and Bcl-2 protein with Kis of 0.5-0.6 μM and 0.2-0.3 mM, respectively. It is harmful if swallowed and suspected of causing cancer. Proper handling and storage are crucial, including avoiding inhalation and contact with eyes and skin, using appropriate ventilation, and storing in a cool, well-ventilated area away from direct sunlight and ignition sources.
  • Binds to Bcl-xL protein and Bcl-2 protein
  • Harmful if swallowed
  • Suspected of causing cancer
  • Avoid inhalation and contact with eyes and skin
  • Use in areas with appropriate exhaust ventilation
  • Keep container tightly sealed in a cool, well-ventilated area
  • Keep away from direct sunlight and sources of ignition