Learn More
Medchemexpress LLC Gossypol (acetic acid) | 12542-36-8 | 99.8% | 578.61 | 5 G
SDP
Catalog No. 5000386570
Encompass_Preferred
Gossypol acetic acid ((±)-Gossypol-acetic acid) binds to Bcl-xL protein and Bcl-2 protein, making it suitable for cancer research and studies involving the Bcl-2 family.
  • Targets Bcl-2 Family
  • Used in cancer research
  • Binds to Bcl-xL protein with a Ki of 0.5-0.6 μM
  • Binds to Bcl-2 protein with a Ki of 0.2-0.3 mM
Catalog No. 50-003-86570 Supplier Medchemexpress LLC Supplier No. HY175105G
Add to Cart
Add to Cart
Gossypol acetic acid ((±)-Gossypol-acetic acid) binds to Bcl-xL protein and Bcl-2 protein, making it suitable for cancer research and studies involving the Bcl-2 family.
  • Targets Bcl-2 Family
  • Used in cancer research
  • Binds to Bcl-xL protein with a Ki of 0.5-0.6 μM
  • Binds to Bcl-2 protein with a Ki of 0.2-0.3 mM