Learn More
Medchemexpress LLC Gossypol (acetic acid) | 12542-36-8 | 99.8% | 578.61 | 50 MG
SDP
Catalog No. 5000386474
Encompass_Preferred
Gossypol acetic acid is a natural product derived from cottonseeds and roots, primarily studied as an anticancer agent. It binds to Bcl-xL protein and Bcl-2 protein, and its racemic form has been evaluated in various clinical trials, demonstrating good tolerability.
  • Binds to Bcl-xL protein with Ki of 0.5-0.6 μM.
  • Binds to Bcl-2 protein with Ki of 0.2-0.3 mM.
  • Exhibits anticancer properties.
  • Tested in several clinical trials.
  • Well tolerated.
Catalog No. 50-003-86474 Supplier Medchemexpress LLC Supplier No. HY1751050MG
Gossypol acetic acid is a natural product derived from cottonseeds and roots, primarily studied as an anticancer agent. It binds to Bcl-xL protein and Bcl-2 protein, and its racemic form has been evaluated in various clinical trials, demonstrating good tolerability.
  • Binds to Bcl-xL protein with Ki of 0.5-0.6 μM.
  • Binds to Bcl-2 protein with Ki of 0.2-0.3 mM.
  • Exhibits anticancer properties.
  • Tested in several clinical trials.
  • Well tolerated.