Learn More
TARGETMOL CHEMICALS INC Gossypol acetic acid 500MG
SDP
Catalog No. 502877600
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Gossypol acetic acid (AT101), a polyphenolic compound isolated from cottonseeds, binds with Bcl-2, Bcl-xL, Mcl-1, and does not inhibit BIR3 domain and BID. Purity 98.57%
Catalog No. 50-287-7600 Supplier TARGETMOL CHEMICALS INC Supplier No. T2730500MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Gossypol acetic acid (AT101), a polyphenolic compound isolated from cottonseeds, binds with Bcl-2, Bcl-xL, Mcl-1, and does not inhibit BIR3 domain and BID. Purity 98.57%