Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-Gossypol-acetic acid, ∼95% (HPLC); ∼95%, MilliporeSigma™ Supelco™

Catalog No. 111012846 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2846 250 mg
11-101-2845 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2846 Supplier MilliporeSigma Supelco Supplier No. G4382250MG
Add to Cart
Add to Cart
Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Glass Bottle
CAS 12542-36-8
Physical Form Neat
Linear Formula C30H30O8 · C2H4O2
MDL Number MFCD00058385
Type Analytical Standard
Quantity 250 mg
Formula Weight 578.61
Percent Purity 95% (HPLC); 95%
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.