Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gossypol-Acetic Acid, MP Biomedicals™

Catalog No. ICN19521125 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19521125 25 mg
ICN19521180 100 mg
ICN19521183 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19521125 Supplier MP Biomedicals, Inc Supplier No. 0219521125
Add to Cart
Add to Cart

Chemical Identifiers

CAS 12542-36-8
Molecular Formula C32H34O10
Molecular Weight (g/mol) 578.614
InChI Key NIOHNDKHQHVLKA-UHFFFAOYSA-N
Synonym gossypol-acetic acid, gossypol acetate, gossypol acetic acid, acetate gossypol, --gossypol acetic acid, s-gossypol acetic acid, at-101 acetic acid, gossypol-acetic acid complex
PubChem CID 227456
IUPAC Name acetic acid;7-(8-formyl-1,6,7-trihydroxy-3-methyl-5-propan-2-ylnaphthalen-2-yl)-2,3,8-trihydroxy-6-methyl-4-propan-2-ylnaphthalene-1-carbaldehyde
SMILES CC1=C(C(=C2C(=C1)C(=C(C(=C2C=O)O)O)C(C)C)O)C3=C(C=C4C(=C3O)C(=C(C(=C4C(C)C)O)O)C=O)C.CC(=O)O

Specifications

Quantity 25 mg
Formula Weight 578.61
Physical Form Solid
Chemical Name or Material Gossypol-Acetic Acid
Recommended Storage Store at 2° to 8°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.