Learn More
Selleck Chemical LLC Gossypol Acetate 100mg 12542-36-8 Gossypol Acetic acid, Pogosin, AT101
SDP
Supplier:  Selleck Chemical LLC S2303100MG
Encompass_Preferred
Gossypol Acetate (Gossypol Acetic acid, Pogosin, AT101) is a polyphenolic aldehyde that permeates cells and acts as an inhibitor for several dehydrogenase enzymes such as lactate dehydrogenase, NAD-linked enzymes. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-808-9