Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gossypol, MP Biomedicals™

Catalog No. ICN19521025 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19521025 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19521025 Supplier MP Biomedicals, Inc Supplier No. 0219521025

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 303-45-7
Molecular Formula C30H30O8
Molecular Weight (g/mol) 518.56
MDL Number MFCD00017352
InChI Key QBKSWRVVCFFDOT-UHFFFAOYSA-N
Synonym gossypol, --gossypol, +-gossypol, pogosin, tash 1, racemic-gossypol, r---gossypol, r-gossypol, +--gossypol, +/--gossypol
PubChem CID 3503
ChEBI CHEBI:28584
IUPAC Name 1,1',6,6',7,7'-hexahydroxy-3,3'-dimethyl-5,5'-bis(propan-2-yl)-[2,2'-binaphthalene]-8,8'-dicarbaldehyde
SMILES CC(C)C1=C(O)C(O)=C(C=O)C2=C(O)C(=C(C)C=C12)C1=C(O)C2=C(C=O)C(O)=C(O)C(C(C)C)=C2C=C1C

Specifications

Color Yellow
Quantity 25 mg
Formula Weight 518.6
Physical Form Solid
Chemical Name or Material Gossypol
Recommended Storage Store at 2° to 8°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.