Learn More
TARGETMOL CHEMICALS INC GOSSYPOLONE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T319875MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Gossypolone is is a proposed major metabolite of gossypol. Gossypolone reduces Notch/Wnt signaling and induces apoptosis. purity: 96%
Catalog No. 50-258-0340