Learn More
Aviva Systems Biology Corp GOT1 Antibody - N-terminal region (ARP48205_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48205_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GOT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GOT1. Alternate names: GIG18, ASTQTL1ApplicationWB. Species reactivity: Cow, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-59758