Learn More
Aviva Systems Biology Corp GOT2 Antibody - C-terminal region (ARP43518_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43518_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GOT2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human GOT2. Alternate names: FLJ40994, kat4, kativ, mitaat, KAT4, KATIV, mitAATApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58558