Learn More
Aviva Systems Biology Corp GOT2 Antibody - N-terminal region (ARP43517_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43517_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GOT2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GOT2. Alternate names: FLJ40994, kat4, kativ, mitaat, KAT4, KATIV, mitAATApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-58557