Learn More
Med Vet International McKesson Chemotherapy Procedure Isolation Gown, 2X Large, Blue, NonSterile, 16-55KV2X, 10/Bag
Catalog No. 503047474
Encompass_Preferred
Chemotherapy Procedure Gown McKesson 2X-Large Blue NonSterile AAMI Level 2 / ASTM D6978 Disposable Polycoated material delivers durability and protection; soft, spunbound polypropylene is coated with a layer of polyethylene film Two back designs: open back or full back Neck closure options: hook-and-loop, over-the-head and tie Secured with ties at waist for added comfort Low-linting material helps to provide a clean environment Long sleeves with knit cuffs provide a comfortable fit Full back gowns are rated AAMI 2
Catalog No. 50-304-7474 Supplier Med Vet International Supplier No. 1177930BG
Chemotherapy Procedure Gown McKesson 2X-Large Blue NonSterile AAMI Level 2 / ASTM D6978 Disposable Polycoated material delivers durability and protection; soft, spunbound polypropylene is coated with a layer of polyethylene film Two back designs: open back or full back Neck closure options: hook-and-loop, over-the-head and tie Secured with ties at waist for added comfort Low-linting material helps to provide a clean environment Long sleeves with knit cuffs provide a comfortable fit Full back gowns are rated AAMI 2