Learn More
Med Vet International McKesson Full Back Chemotherapy Procedure Gown, Medium, Blue, NonSterile, 16-54KVM, 10/Bag
Catalog No. 503047478
Encompass_Preferred
Chemotherapy Procedure Gown McKesson Medium Blue NonSterile AAMI Level 2 / ASTM D6978 Disposable Medium size, 41 inches Made of soft durable material for comfort and quality Secures with ties at the waist for added comfort Long sleeves with knit cuffs provide a comfortable fit Low-linting material helps to provide a clean environment
Catalog No. 50-304-7478 Supplier Med Vet International Supplier No. 1177927BG
Chemotherapy Procedure Gown McKesson Medium Blue NonSterile AAMI Level 2 / ASTM D6978 Disposable Medium size, 41 inches Made of soft durable material for comfort and quality Secures with ties at the waist for added comfort Long sleeves with knit cuffs provide a comfortable fit Low-linting material helps to provide a clean environment