Learn More
Med Vet International McKesson Full Back Chemotherapy Procedure Gown, Extra Large, Blue, NonSterile, 16-55KVX, 10/Bag
Catalog No. 503047500
Encompass_Preferred
Chemotherapy Procedure Gown McKesson X-Large Blue NonSterile AAMI Level 2 / ASTM D6978 Disposable X-Large size, 51 inches Made of soft durable material for comfort and quality Secures with ties at the waist for added comfort Long sleeves with knit cuffs provide a comfortable fit Low-linting material helps to provide a clean environment
Catalog No. 50-304-7500 Supplier Med Vet International Supplier No. 1177929BG
Chemotherapy Procedure Gown McKesson X-Large Blue NonSterile AAMI Level 2 / ASTM D6978 Disposable X-Large size, 51 inches Made of soft durable material for comfort and quality Secures with ties at the waist for added comfort Long sleeves with knit cuffs provide a comfortable fit Low-linting material helps to provide a clean environment