Learn More
Med Vet International Patient Exam Gown X-Large Blue Disposable, 70260N, 25/Case
Catalog No. 502831310
Encompass_Preferred
Tissue/poly gown consists of tissue bonded to a fluid-resistant layer of polyethylene that will absorb fluid on one side and protect on the other Reversible with a front or back opening
Catalog No. 50-283-1310 Supplier Med Vet International Supplier No. 203271CS
Tissue/poly gown consists of tissue bonded to a fluid-resistant layer of polyethylene that will absorb fluid on one side and protect on the other Reversible with a front or back opening