Learn More
Med Vet International McKesson Patient Exam Gown, Teal, One Size Fits Most, Adult, Unisex, 18-973, 50/Case
Catalog No. 503047466
Encompass_Preferred
Patient Exam Gown McKesson One Size Fits Most Teal Disposable Premium Tissue/Poly/Tissue 30 X 42 inch, Teal Waist Belt 56-inch sweep Provides knee-length privacy and helps protect against fluid strike-through Front/back opening Not made with natural rubber latex
Catalog No. 50-304-7466 Supplier Med Vet International Supplier No. 224226CS
Patient Exam Gown McKesson One Size Fits Most Teal Disposable Premium Tissue/Poly/Tissue 30 X 42 inch, Teal Waist Belt 56-inch sweep Provides knee-length privacy and helps protect against fluid strike-through Front/back opening Not made with natural rubber latex