Learn More
Med Vet International ProtectIVe Procedure Gown Precept One Size Fits Most Yellow Nonsterile Aami Level 2 Disposable, 51177, 10/Bag, 10 Bags
Catalog No. 502832120
Encompass_Preferred
Size: Large (length 45 inches, width 55 inches)Secures with neck ties Elastic wrist cuffs Sonic welded seams Full coverage with overlapping back panels
Catalog No. 50-283-2120 Supplier Med Vet International Supplier No. 1197109CS
Size: Large (length 45 inches, width 55 inches)Secures with neck ties Elastic wrist cuffs Sonic welded seams Full coverage with overlapping back panels