Learn More
Med Vet International Dynarex CPE Thumb Loop Isolation Gown, Universal, Yellow, 50/Case
Catalog No. 503047489
Encompass_Preferred
Dynarex Isolation Gowns are professional-grade, disposable garments designed to provide effective protection from germs and cross-contamination. These full-coverage gowns are constructed from lightweight, breathable material and feature sewn seams and extra-long waist ties that can be secured in the front of the body. Constructed from non-woven material Gowns feature elastic cuffs, sewn seams, and extra-long waist ties Full-coverage and universally sized Individually wrapped Disposable Fluid-resistant (2141) and impervious, poly-coated material (2144-2148) available CPE/Thumb Loop (2140) comes in a perforated box for convenient dispensing Not made with natural rubber latex
Catalog No. 50-304-7489 Supplier Med Vet International Supplier No. 2141
Dynarex Isolation Gowns are professional-grade, disposable garments designed to provide effective protection from germs and cross-contamination. These full-coverage gowns are constructed from lightweight, breathable material and feature sewn seams and extra-long waist ties that can be secured in the front of the body. Constructed from non-woven material Gowns feature elastic cuffs, sewn seams, and extra-long waist ties Full-coverage and universally sized Individually wrapped Disposable Fluid-resistant (2141) and impervious, poly-coated material (2144-2148) available CPE/Thumb Loop (2140) comes in a perforated box for convenient dispensing Not made with natural rubber latex