Learn More
Med Vet International ProtectIVe Procedure Gown Cardinal Health One Size Fits Most Yellow Nonsterile Not Rated Disposable, 1100Pg, 10/Pack, 10 Packs
Catalog No. 502832465
Encompass_Preferred
Basic spunbond polypropylene isolation gowns Comfort and protection for use in very minimal fluid settings Generously sized for greater coverage and flexibility
Catalog No. 50-283-2465 Supplier Med Vet International Supplier No. 466476CS
Basic spunbond polypropylene isolation gowns Comfort and protection for use in very minimal fluid settings Generously sized for greater coverage and flexibility