Learn More
Med Vet International Quickcomply ProtectIVe Procedure Gown, X-Large, Yellow, Nonsterile, Aami Level 2, Disposable, Nonlv240Xl, 10/Bag
Catalog No. 502830760
Encompass_Preferred
Quick Comply gowns are designed to reduce donning and removal time Over-the-head style eliminates closing the neck Made from a multilayer nonwoven material that is AAMI PB70 level 2 compliant Strong sealed seams help eliminate strike through under light to moderate fluid pressure conditions Generously sized gown features thumb loop wrists and side ties, and it provides full back coverage Sealed Seams
Catalog No. 50-283-0760 Supplier Med Vet International Supplier No. 1108657BG
Quick Comply gowns are designed to reduce donning and removal time Over-the-head style eliminates closing the neck Made from a multilayer nonwoven material that is AAMI PB70 level 2 compliant Strong sealed seams help eliminate strike through under light to moderate fluid pressure conditions Generously sized gown features thumb loop wrists and side ties, and it provides full back coverage Sealed Seams