Learn More
Med Vet International McKesson Open Back Over-the-Head Protective Procedure Gown, Universal, Blue, 18-8576A, 75/Case
Catalog No. 503047476
Encompass_Preferred
Protective Procedure Gown McKesson One Size Fits Most Blue NonSterile AAMI Level 2 Disposable AAMI Level 2 Rated An open back allows for increased airflow and comfort One size fits most
Catalog No. 50-304-7476 Supplier Med Vet International Supplier No. 1172304CS
Protective Procedure Gown McKesson One Size Fits Most Blue NonSterile AAMI Level 2 Disposable AAMI Level 2 Rated An open back allows for increased airflow and comfort One size fits most