Learn More
Med Vet International McKesson Non-Reinforced Surgical Gown with Towel, X-Large, Blue, Sterile, 183-I90-8030-S1, 1/Pack
Catalog No. 503047444
Encompass_Preferred
Non-Reinforced Surgical Gown with Towel McKesson X-Large Blue Sterile AAMI Level 3 Disposable Size x-large with 52 Inch length Blue AAMI Level 3 Provides moderate-to-high level protection for operating room personnel and surgical patients from the transfer of microorganisms, bodily fluids and particulate Triple-density, SMS material provides excellent barrier strength, protection and breathability Intuitive rear pleats, generous cuts and raglan-sleeve design provide increased range of motion and enhanced comfort Extra-long hook-and-loop feature creates an adjustable fit around the neckline, aids in donning, and maximizes comfort Set-in sleeve designs with knit cuffs feature generous cuts and intuitive rear pleats for enhanced comfort and movability Included transfer tag and waist ties for easy donning and notetaking for convenience in the sterile field
Catalog No. 50-304-7444 Supplier Med Vet International Supplier No. 1104453PK
 Restricted Item
Add to Cart
Add to Cart
Non-Reinforced Surgical Gown with Towel McKesson X-Large Blue Sterile AAMI Level 3 Disposable Size x-large with 52 Inch length Blue AAMI Level 3 Provides moderate-to-high level protection for operating room personnel and surgical patients from the transfer of microorganisms, bodily fluids and particulate Triple-density, SMS material provides excellent barrier strength, protection and breathability Intuitive rear pleats, generous cuts and raglan-sleeve design provide increased range of motion and enhanced comfort Extra-long hook-and-loop feature creates an adjustable fit around the neckline, aids in donning, and maximizes comfort Set-in sleeve designs with knit cuffs feature generous cuts and intuitive rear pleats for enhanced comfort and movability Included transfer tag and waist ties for easy donning and notetaking for convenience in the sterile field