Learn More
Med Vet International McKesson Non-Reinforced Surgical Gown with Towel, Medium, Blue, Sterile, 183-I90-8010-S1, 32/Case
Catalog No. 503047439
Encompass_Preferred
Non-Reinforced Surgical Gown with Towel McKesson Medium Blue Sterile AAMI Level 3 Disposable Size medium with 45 Inch length Blue AAMI Level 3 Triple-density, SMS material provides excellent barrier strength, protection and breathability Intuitive rear pleats, generous cuts and raglan-sleeve design provide increased range of motion and enhanced comfort Extra-long hook-and-loop feature creates an adjustable fit around the neckline, aids in donning, and maximizes comfort Set-in sleeve designs with knit cuffs feature generous cuts and intuitive rear pleats for enhanced comfort and movability Included transfer tag and waist ties for easy donning and notetaking for convenience in the sterile field
Catalog No. 50-304-7439 Supplier Med Vet International Supplier No. 1104451CS
Non-Reinforced Surgical Gown with Towel McKesson Medium Blue Sterile AAMI Level 3 Disposable Size medium with 45 Inch length Blue AAMI Level 3 Triple-density, SMS material provides excellent barrier strength, protection and breathability Intuitive rear pleats, generous cuts and raglan-sleeve design provide increased range of motion and enhanced comfort Extra-long hook-and-loop feature creates an adjustable fit around the neckline, aids in donning, and maximizes comfort Set-in sleeve designs with knit cuffs feature generous cuts and intuitive rear pleats for enhanced comfort and movability Included transfer tag and waist ties for easy donning and notetaking for convenience in the sterile field