Learn More
Med Vet International Level 2 Eclipse Disposable Surgical Gown, Sterile, With Towel, Sms, Large, 30 Count
Catalog No. 502829328
Encompass_Preferred
A Traditional Favorite, Medline's Eclipse Line Is Extremely Appealing Due To Its Cloth-Like Texture And Softness. For Many Customers, Surgical Gowns Are Daily Attire, So A Truly Comfortable Gown Is Appreciated And Readily Worn. DuPont's Softesse Material Is Largely Biodegradable, Being Composed Of Both Wood Pulp And Polyester--So Even The Surgical World Can Be Green. This Fabric's Advanced Splash-Resistant Barrier Gives You Both Breathability And Great Protection. Dynjp2001H: Eclipse, L-43" And Non-Reinforced. AAMI Level 2 protection
Catalog No. 50-282-9328 Supplier Med Vet International Supplier No. DYNJP2001
A Traditional Favorite, Medline's Eclipse Line Is Extremely Appealing Due To Its Cloth-Like Texture And Softness. For Many Customers, Surgical Gowns Are Daily Attire, So A Truly Comfortable Gown Is Appreciated And Readily Worn. DuPont's Softesse Material Is Largely Biodegradable, Being Composed Of Both Wood Pulp And Polyester--So Even The Surgical World Can Be Green. This Fabric's Advanced Splash-Resistant Barrier Gives You Both Breathability And Great Protection. Dynjp2001H: Eclipse, L-43" And Non-Reinforced. AAMI Level 2 protection