Learn More
Med Vet International McKesson Fabric Reinforced Surgical Gown with Towel, Large, Blue, Sterile, 183-I91-4320-S1, 1/Pack
Catalog No. 503047473
Encompass_Preferred
Fabric-Reinforced Surgical Gown with Towel McKesson Large Blue Sterile AAMI Level 3 Disposable Size large with 50 Inch length Blue AAMI Level 3 Provides moderate-to-high level protection for operating room personnel and surgical patients from the transfer of microorganisms, bodily fluids and particulate Triple-density, SMS material provides excellent barrier strength, protection and breathability Intuitive rear pleats, generous cuts and raglan-sleeve design provide increased range of motion and enhanced comfort Extra-long hook-and-loop feature creates an adjustable fit around the neckline, aids in donning, and maximizes comfort Surgical-fit cuffs feature extended lengths and unique construction to ensure a better fit and softer feel Included transfer tag and waist ties for easy donning and notetaking for convenience in the sterile field
Catalog No. 50-304-7473 Supplier Med Vet International Supplier No. 1101271PK
Fabric-Reinforced Surgical Gown with Towel McKesson Large Blue Sterile AAMI Level 3 Disposable Size large with 50 Inch length Blue AAMI Level 3 Provides moderate-to-high level protection for operating room personnel and surgical patients from the transfer of microorganisms, bodily fluids and particulate Triple-density, SMS material provides excellent barrier strength, protection and breathability Intuitive rear pleats, generous cuts and raglan-sleeve design provide increased range of motion and enhanced comfort Extra-long hook-and-loop feature creates an adjustable fit around the neckline, aids in donning, and maximizes comfort Surgical-fit cuffs feature extended lengths and unique construction to ensure a better fit and softer feel Included transfer tag and waist ties for easy donning and notetaking for convenience in the sterile field