Learn More
Andwin Scientific VWR Irradiated Sleeves, Sterile, Microporous, White, Universal, 19½", 3¼" (relaxed cuff), 3½" (stretched cuff) / 4½" (relaxed arm), 9¾" (stretched arm)
SDP
Catalog No. NC2262563
Encompass_Preferred
Reliable sterile sleeves for arm coverage in cleanroom and controlled-environment workflows, using breathable low-linting fabric that meets ISO and GMP requirements for sterile gowning.
Gamma irradiated to a Sterility Assurance Level (SAL) of 10⁻⁶ to support sterile cleanroom gowning workflows
Breathable microporous construction balances barrier performance with comfort during extended wear
Low particle-shedding material helps reduce contamination risk in controlled processing areas
Elastic openings help maintain sleeve position through repetitive reaching and handling
Available with or without thumb loops to match gowning preferences
Designed for critical gowning workflows, these sleeves add focused arm coverage in controlled environments where contamination control matters. The microporous construction helps balance barrier performance with breathability for comfortable wear through routine movement.
This item is not returnable. View return policy
Catalog No. NC2262563 Supplier Andwin Scientific Supplier No. 414004510
Reliable sterile sleeves for arm coverage in cleanroom and controlled-environment workflows, using breathable low-linting fabric that meets ISO and GMP requirements for sterile gowning.
Gamma irradiated to a Sterility Assurance Level (SAL) of 10⁻⁶ to support sterile cleanroom gowning workflows
Breathable microporous construction balances barrier performance with comfort during extended wear
Low particle-shedding material helps reduce contamination risk in controlled processing areas
Elastic openings help maintain sleeve position through repetitive reaching and handling
Available with or without thumb loops to match gowning preferences
Designed for critical gowning workflows, these sleeves add focused arm coverage in controlled environments where contamination control matters. The microporous construction helps balance barrier performance with breathability for comfortable wear through routine movement.