Learn More
Med Vet International Level 3 Sirus Disposable Surgical Gown, Sterile, Sms, 2X-Large, 18 Count
Catalog No. 502092213
Encompass_Preferred
Medline's Sirus line offers an incredibly economical gown without sacrificing comfort or barrier strength. Our SMS fabric binds three layers of fabric--a densely-packed meltblown layer sandwiched between two, strong spunbond outer layers--to provide incredible fluid repellency. SMS is the new industry standard, but ours is even a little better. Progressive Reicofil technology offers customers increased fabric uniformity, equipping the clinician with the market's latest-generation of SMS. DYNJP2003S: Sirus, XXL-49" and non reinforced.Level 3
Catalog No. 50-209-2213 Supplier Med Vet International Supplier No. DYNJP2003S
Medline's Sirus line offers an incredibly economical gown without sacrificing comfort or barrier strength. Our SMS fabric binds three layers of fabric--a densely-packed meltblown layer sandwiched between two, strong spunbond outer layers--to provide incredible fluid repellency. SMS is the new industry standard, but ours is even a little better. Progressive Reicofil technology offers customers increased fabric uniformity, equipping the clinician with the market's latest-generation of SMS. DYNJP2003S: Sirus, XXL-49" and non reinforced.Level 3