Learn More
Med Vet International Dynarex Sterile Surgical Gowns Reinforced, Medium, 20/Case
Catalog No. 503047459
Encompass_Preferred
Impervious to blood, liquids and other potentially infectious material. ANSI/AAMI PB70, Level 3 Protection Poly-reinforcement repels fluids Low linting fabric Wrap protects surgical gown and provides sterilized field Ties in back Contents: (1)Gown, breathable; (2)Hand towels; (1)Wrap
Catalog No. 50-304-7459 Supplier Med Vet International Supplier No. 8192
Impervious to blood, liquids and other potentially infectious material. ANSI/AAMI PB70, Level 3 Protection Poly-reinforcement repels fluids Low linting fabric Wrap protects surgical gown and provides sterilized field Ties in back Contents: (1)Gown, breathable; (2)Hand towels; (1)Wrap