Learn More
Med Vet International Dynarex Sterile Surgical Gowns Reinforced, X-Large, 20/Case
Catalog No. 503047480
Encompass_Preferred
Impervious to blood, liquids and other potentially infectious material. ANSI/AAMI PB70, Level 3 Protection Poly-reinforcement repels fluids Low linting fabric Wrap protects surgical gown and provides sterilized field Ties in back Contents: (1)Gown, Breathable; (2)Hand Towels; (1)Wrap
Catalog No. 50-304-7480 Supplier Med Vet International Supplier No. 8194
 Restricted Item
Add to Cart
Add to Cart
Impervious to blood, liquids and other potentially infectious material. ANSI/AAMI PB70, Level 3 Protection Poly-reinforcement repels fluids Low linting fabric Wrap protects surgical gown and provides sterilized field Ties in back Contents: (1)Gown, Breathable; (2)Hand Towels; (1)Wrap