Learn More
Medchemexpress LLC Gozanertinib | 1226549-49-0 | 98.4% | 10 MG
SDP
Catalog No. 5000340060
Encompass_Preferred
Gozanertinib is an orally active furanopyrimidine-based EGFR inhibitor. It occupies the ATP-binding site and demonstrates significant antitumor efficacy by inhibiting both wild-type EGFR and the L858R/T790M mutant. This compound is for research use only.
  • Orally active furanopyrimidine-based EGFR inhibitor
  • Occupies the ATP-binding site
  • Inhibits EGFRWT with an IC50 of 15 nM
  • Inhibits EGFRL858R/T790M with an IC50 of 48 nM
  • Demonstrates significant antitumor efficacy
  • Interacts via covalent bonding, hydrogen bonds, and hydrophobic interactions
  • Shows inhibitory activity against HCC827, H1975, and A431 cell lines
Catalog No. 50-003-40060 Supplier Medchemexpress LLC Supplier No. HY12877810MG
Gozanertinib is an orally active furanopyrimidine-based EGFR inhibitor. It occupies the ATP-binding site and demonstrates significant antitumor efficacy by inhibiting both wild-type EGFR and the L858R/T790M mutant. This compound is for research use only.
  • Orally active furanopyrimidine-based EGFR inhibitor
  • Occupies the ATP-binding site
  • Inhibits EGFRWT with an IC50 of 15 nM
  • Inhibits EGFRL858R/T790M with an IC50 of 48 nM
  • Demonstrates significant antitumor efficacy
  • Interacts via covalent bonding, hydrogen bonds, and hydrophobic interactions
  • Shows inhibitory activity against HCC827, H1975, and A431 cell lines