Learn More
Medchemexpress LLC Gozanertinib | 1226549-49-0 | 98.4% | 100 MG
SDP
Catalog No. 5000338580
Encompass_Preferred
Gozanertinib, also known as DBPR112, is an orally active furanopyrimidine-based EGFR inhibitor. It functions by occupying the ATP-binding site and demonstrates significant antitumor efficacy.
  • Inhibits EGFRWT with an IC50 of 15 nM
  • Inhibits EGFRL858R/T790M with an IC50 of 48 nM
  • Induces dose-dependent reduction of phosphorylated EGFR in H1975 cells
  • Demonstrates inhibitory activity against HCC827 (CC50=25 nM), H1975 (CC50=620 nM), and A431 (CC50=1.02 μM) cell lines
  • Reduces tumor growth in HCC827 tumor models
  • Shows antitumor effect in H1975 tumor models
Catalog No. 50-003-38580 Supplier Medchemexpress LLC Supplier No. HY128778100MG
Gozanertinib, also known as DBPR112, is an orally active furanopyrimidine-based EGFR inhibitor. It functions by occupying the ATP-binding site and demonstrates significant antitumor efficacy.
  • Inhibits EGFRWT with an IC50 of 15 nM
  • Inhibits EGFRL858R/T790M with an IC50 of 48 nM
  • Induces dose-dependent reduction of phosphorylated EGFR in H1975 cells
  • Demonstrates inhibitory activity against HCC827 (CC50=25 nM), H1975 (CC50=620 nM), and A431 (CC50=1.02 μM) cell lines
  • Reduces tumor growth in HCC827 tumor models
  • Shows antitumor effect in H1975 tumor models