Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gozanertinib | 1226549-49-0 | 98.4% | 50 MG
SDP

Catalog No. 5000341344
Encompass_Preferred

Gozanertinib is an orally active furanopyrimidine-based EGFR inhibitor with IC50s of 15 nM for EGFRWT and 48 nM for EGFRL858R/T790M. It can occupy the ATP-binding site and demonstrates significant antitumor efficacy. This compound is intended for research use only.

  • Orally active EGFR inhibitor
  • IC50 of 15 nM for EGFRWT
  • IC50 of 48 nM for EGFRL858R/T790M
  • Occupies ATP-binding site
  • Demonstrates significant antitumor efficacy
  • Molecular weight: 533.62
  • Formula: C32H31N5O3
  • Appearance: solid, off-white to light yellow
  • Inhibits HCC827, H1975, and A431 cell lines
  • Reduces phosphorylated EGFR in a dose-dependent manner

Catalog No. 50-003-41344 Supplier Medchemexpress LLC Supplier No. HY12877850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gozanertinib is an orally active furanopyrimidine-based EGFR inhibitor with IC50s of 15 nM for EGFRWT and 48 nM for EGFRL858R/T790M. It can occupy the ATP-binding site and demonstrates significant antitumor efficacy. This compound is intended for research use only.

  • Orally active EGFR inhibitor
  • IC50 of 15 nM for EGFRWT
  • IC50 of 48 nM for EGFRL858R/T790M
  • Occupies ATP-binding site
  • Demonstrates significant antitumor efficacy
  • Molecular weight: 533.62
  • Formula: C32H31N5O3
  • Appearance: solid, off-white to light yellow
  • Inhibits HCC827, H1975, and A431 cell lines
  • Reduces phosphorylated EGFR in a dose-dependent manner

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.