Learn More
Gerstel GP-2M ADAPTER FOR OD .45MM COL
SDP
Catalog No. NC9596323
Encompass
CIS column adapter for 0.25 mm and 0.32 mm ID capillary columns. Use with 001805-040-00 (0.25 mm ID) or 001805-045-00 (0.32 mm ID) GP-2M ferrules.
Catalog No. NC9596323 Supplier Gerstel Supplier No. 00725904500
CIS column adapter for 0.25 mm and 0.32 mm ID capillary columns. Use with 001805-040-00 (0.25 mm ID) or 001805-045-00 (0.32 mm ID) GP-2M ferrules.