Learn More
MSC Momentive Performance Materials IS808 12C 10.1 oz Tube Clear RTV Silicone Joint Sealant -50 to 200°C Operating Temp, 25 min Tack Free Dry Time, 24 hr Full Cure Time, Series IS800
Catalog No. 501113824
Encompass
Momentive Performance Materials IS808 12C 10.1 oz Tube Clear RTV Silicone Joint Sealant -50 to 200°C Operating Temp, 25 min Tack Free Dry Time, 24 hr Full Cure Time, Series IS800
Catalog No. 50-111-3824 Supplier MSC Supplier No. 06904700
Momentive Performance Materials IS808 12C 10.1 oz Tube Clear RTV Silicone Joint Sealant -50 to 200°C Operating Temp, 25 min Tack Free Dry Time, 24 hr Full Cure Time, Series IS800