Learn More
Cayman Chemical GP-AMChydrobromIde 50mg
SDP
Catalog No. 502946578
Encompass_Preferred
A fluorogenic substrate for DPP-4; AMC is released upon enzymatic cleavage by DPP-4 and its fluorescence can be used to quantify DPP-4 activity; ex/em = 340-360/440-460 nm, respectively
Catalog No. 50-294-6578 Supplier Cayman Chemical Supplier No. 3445850 MG
A fluorogenic substrate for DPP-4; AMC is released upon enzymatic cleavage by DPP-4 and its fluorescence can be used to quantify DPP-4 activity; ex/em = 340-360/440-460 nm, respectively