Learn More
Cayman Chemical GP-AMChydrobromIde 5mg
SDP
Catalog No. 502949561
Encompass_Preferred
A fluorogenic substrate for DPP-4; AMC is released upon enzymatic cleavage by DPP-4 and its fluorescence can be used to quantify DPP-4 activity; ex/em = 340-360/440-460 nm, respectively
Catalog No. 50-294-9561 Supplier Cayman Chemical Supplier No. 344585 MG
A fluorogenic substrate for DPP-4; AMC is released upon enzymatic cleavage by DPP-4 and its fluorescence can be used to quantify DPP-4 activity; ex/em = 340-360/440-460 nm, respectively