Learn More
Enzo Life Sciences GP Antagonist 2 (1mg)
SDP
Supplier:  Enzo Life Sciences BMLG5100001
Encompass_Preferred
Selective, competitive and reversible G-protein antagonist. Inhibits the activation of Gi or Go by M2 muscarinic cholinergic receptors (mAChR) and Gs by β-adrenergic receptors in reconstituted phospholipid vesicles. Inhibition is reversible and competitive with respect to receptor binding to G proteins. Substance P (ALX-167-003) related compound. Alternative name: G protein antagonist 2. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water or DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-0886