Learn More
DIGI KEY CORPORATION General purpose dGP ust remover 14oz, non-flammable
SDP
Catalog No. NC3684663
Encompass
MicroCare's General Purpose Dust Remover (MCC-DST14A) is packed in a 14 oz aerosol can that delivers 70-140 psi to remove dust from electronic equipment. This dust remover spray for electronics is non-flammable, making it safe to use on energized equipment.
Catalog No. NC3684663 Supplier DIGI KEY CORPORATION Supplier No. 2805MCCDST14AND
MicroCare's General Purpose Dust Remover (MCC-DST14A) is packed in a 14 oz aerosol can that delivers 70-140 psi to remove dust from electronic equipment. This dust remover spray for electronics is non-flammable, making it safe to use on energized equipment.