Learn More
Cole-Parmer Digi-Sense Gen-Purp Fle x Thermistor, Vinyl Sheath and Tip, Phono Plug, 50ft Lead
Catalog No. 501961495 Shop All Cole-Parmer Products
Encompass_Preferred
Thermistor probes offer excellent accuracy over the biological temperature range. Probes are electronically isolated and come with a 10 ft. lead with a phono plug (unless otherwise specified). Probes are designed for multiple applications. Time Constant: 1-4 seconds or greater
Catalog No. 50-196-1495 Supplier Cole-Parmer Supplier No. 0849104
Thermistor probes offer excellent accuracy over the biological temperature range. Probes are electronically isolated and come with a 10 ft. lead with a phono plug (unless otherwise specified). Probes are designed for multiple applications. Time Constant: 1-4 seconds or greater