Learn More
MSC Diversey General Purpose Cleaner, Concentrated, Citrus, 1.5 L, 2/Carton
Catalog No. 502836358
Encompass
This highly versatile general purpose cleaner concentrate is designed to effectively clean a variety of water-safe surfaces. It contains high-performance detergents that remove dirt, grit, grease, and soap scum without rinsing. Featuring a pleasant citrus scent, it is suitable for use in mop and bucket, auto-scrubber, and trigger spray applications.
  • Effectively cleans counters, tables, floors, and other hard surfaces.
  • Concentrated formulation with a 1:256 dilution ratio.
  • Removes dirt, grit, grease, and soap scum without requiring rinsing.
  • Features a pleasant citrus scent.
  • Compatible with cold, hot, soft, and hard water.
  • Suitable for mop and bucket, auto-scrubber, and trigger spray applications.
  • OU Kosher certified.
Catalog No. 50-283-6358 Supplier MSC Supplier No. 42581157
This highly versatile general purpose cleaner concentrate is designed to effectively clean a variety of water-safe surfaces. It contains high-performance detergents that remove dirt, grit, grease, and soap scum without rinsing. Featuring a pleasant citrus scent, it is suitable for use in mop and bucket, auto-scrubber, and trigger spray applications.
  • Effectively cleans counters, tables, floors, and other hard surfaces.
  • Concentrated formulation with a 1:256 dilution ratio.
  • Removes dirt, grit, grease, and soap scum without requiring rinsing.
  • Features a pleasant citrus scent.
  • Compatible with cold, hot, soft, and hard water.
  • Suitable for mop and bucket, auto-scrubber, and trigger spray applications.
  • OU Kosher certified.