Learn More
Enzo Life Sciences Glycogen phosphorylase inhibitor (1mg). CAS: 648926-15-2
SDP
Catalog No. 502013385
Encompass_Preferred
Alternative name: 1-(3-(3-(2-Chloro-4,5-difluorobenzoyl)ureido)-4-methoxyphenyl)-3-methylurea. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.
Catalog No. 50-201-3385 Supplier Enzo Life Sciences Supplier No. ENZCHM1100001
Alternative name: 1-(3-(3-(2-Chloro-4,5-difluorobenzoyl)ureido)-4-methoxyphenyl)-3-methylurea. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.