Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger GP Motor 1/4 HP 1 725 RPM 115V

Catalog No. 502793106
Encompass_Preferred

GP Motor 1/4 HP 1 725 RPM 115V AC 48

Catalog No. 50-279-3106 Supplier Grainger Supplier No. 19YW03
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GP Motor 1/4 HP 1 725 RPM 115V AC 48

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.